missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-8 Swine ELISA Kit DFS Item

Cytokine chemoattractant protein for neutrophils

$538.00 - $998.00

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 3576
Precision <10pg/mL
Regulatory Status RUO - research use only
Sample Size 50μL, 100μL supernatant
View More Specs

Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KSC0081 View Documents Invitrogen™
LSKSC0081
96 assays Each for $538.00
KSC0082 View Documents Invitrogen™
LSKSC0082
192 assays Each for $998.00
Description

Description

The IL-8 Swine ELISA Kit quantifies natural and recombinant swine IL-8 in serum, buffered solution, or cell culture medium.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

Specifications

Specifications

Colorimetric
<10pg/mL
50μL, 100μL supernatant
Interleukin 8
3576
RUO - research use only
Serum, Supernatants
4 hrs

For Research Use Only. Not for use in diagnostic procedures.