Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-8 Human ELISA Kit DFS Item


Designed to detect and quantify the level of human IL-8 in serum, plasma, and supernatant using 96-well plates and a microplate reader

Manufacturer: invitrogen™  LSKHC0082

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHC0082

  • / Each
 

Description & Specifications

Specifications

Test Time 2.5 hrs
Target Interleukin 8
For Use With (Application) Colorimetric
Gene ID (Entrez) 3576
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Precision <5pg/mL
Quantity 192 assays
Sample Type Plasma, Serum, Supernatants
Regulatory Status RUO - research use only
Sample Size 50μL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.