missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-6 Rat ELISA Kit DFS Item

Cytokine with actions in immune response

$420.80 - $1,462.65

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 24498
Precision <5pg/mL
Regulatory Status RUO - research use only
Sample Size 50.0μL
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, detector antibody, anti-IgG-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KRC0061C Invitrogen™
LSKRC0061C
480 assays Each for $1,462.65
KRC0062 Invitrogen™
LSKRC0062
192 assays Each for $980.70
KRC0061 Invitrogen™
LSKRC0061
96 assays Each for $420.80
Specifications

Description

The IL-6 Rat ELISA Kit quantifies natural and recombinant rat IL-6 in serum, buffered solution, or cell culture medium.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Colorimetric
<5pg/mL
50.0μL
Interleukin 6
24498
RUO - research use only
Serum, Supernatants
4.5 hrs