Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-6 Rat ELISA Kit DFS Item


Cytokine with actions in immune response

Manufacturer: invitrogen™  LSKRC0061C

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KRC0061C

  • / Each
 

Description & Specifications

Specifications

Accession Number NM_012589.1, NP_036721.1
Detection Method Colorimetric
Concentration 23.5 to 1500pg/mL
Description IL-6 ELISA Kit, 480 Assays, 23.5 to 1500pg/mL Concentration, 50.0μL Sample Volume, <5pg/mL Sensitivity, ILg6, Ifnb2 Gene Aliases, NM_012589.1, NP_036721.1 Accession Number, 24498 Gene ID, IL6 Gene Symbol, HRP Label, Rat Species, Interleukin 6 Target
Format Liquid
Formulation Liquid
For Use With (Application) Colorimetric, Protein assays and analysis
Gene ID (Entrez) 24498
Includes Pre-coated plate, standard, detector antibody, anti-IgG-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Incubation Time 4.5 hr.
For Use With (Equipment) Microplate reader
Kit Contents Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s) and stop solution
Marker Gene IL6
Product Type IL-6 ELISA Kit
Packaging Quantity 1 Each
Precision <5pg/mL
Quantity 480 assays
Regulatory Status RUO - research use only
Sensitivity <5pg/mL
Sample Type Serum, Supernatants
Sample Size 50.0μL
Sample Volume 50μL
Size 480 Assay(s)
Species Rat
Storage Requirements 2 to 8°C
Test Time 4.5 hrs
Target Interleukin 6
Protein Family Cytokines and receptors
Research Discipline Immunology
Gene Symbols IL6

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.