Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-4 Swine ELISA Kit DFS Item


Key cytokine in the immune network

Manufacturer: invitrogen™  LSKSC0041

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KSC0041

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Quantity 96 assays
Sample Type Serum, Supernatants
Precision 2pg/mL
Sample Size 50μL
For Use With (Application) Colorimetric
Regulatory Status RUO - research use only
Test Time 3 hrs
Target Interleukin 4
Gene ID (Entrez) 397225

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.