Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-4 Human Ultrasensitive ELISA Kit DFS Item


Manufacturer: invitrogen™  LSKHC0044

 View more versions of this product

Catalog No. KHC0044

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Sample Type Plasma, Serum, Supernatants
Sample Size 100μL
Precision <0.27pg/mL
Regulatory Status RUO - research use only
For Use With (Application) Colorimetric
Quantity 96 assays
Gene ID (Entrez) 3565
Target Interleukin 4
Test Time 3.5 hrs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.