Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-4 Human Ultrasensitive ELISA Kit DFS Item


Manufacturer: invitrogen™  LSKHC0043

 View more versions of this product

Catalog No. KHC0043

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Precision <0.27pg/mL
For Use With (Application) Colorimetric
Gene ID (Entrez) 3565
Quantity 192 assays
Regulatory Status RUO - research use only
Sample Type Plasma, Serum, Supernatants
Sample Size 100μL
Test Time 3.5 hrs
Target Interleukin 4

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.