Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-4 Human Ultrasensitive ELISA Kit DFS Item

$701.40 - $1,192.80

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 3565
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Precision <0.27pg/mL
Regulatory Status RUO - research use only
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

KHC0043

DFS Item
 
invitrogen™
LSKHC0043
192 assays Each for $1,192.80

KHC0044

DFS Item
 
invitrogen™
LSKHC0044
96 assays Each for $701.40
Description & Specifications

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 3565
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Precision <0.27pg/mL
Regulatory Status RUO - research use only
Sample Size 100μL
Sample Type Plasma, Serum, Supernatants
Target Interleukin 4
Test Time 3.5 hrs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.