Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-2 Rat ELISA Kit DFS Item


Promotes T-cell growth

Manufacturer: invitrogen™  LSKRC0021

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KRC0021

  • / Each
 

Description & Specifications

Specifications

Precision <5pg/mL
Sample Type Serum, Supernatants
Sample Size 50μL
Target Interleukin 2
Gene ID (Entrez) 116562
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Test Time 3 hrs
For Use With (Application) Colorimetric
Regulatory Status RUO - research use only
Quantity 96 assays

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.