missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-2 Mouse ELISA Kit DFS Item

Designed to detect and quantify the level of mouse IL-2 in serum, plasma, and supernatant using 96-well plates and a microplate reader

$420.80 - $1,462.65

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 16183
Precision <8pg/mL
Regulatory Status RUO - research use only
Sample Size 50μL
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KMC0021 Invitrogen™
LSKMC0021
96 assays Each for $420.80
KMC0021C Invitrogen™
LSKMC0021C
480 assays Each for $1,462.65
KMC0022 Invitrogen™
LSKMC0022
192 assays Each for $980.70
Specifications

Description

Easy to run
Rapid protocol
Convenient precoated, removable 8-well strips

Easy-to-Run Sandwich ELISA
The total assay incubation time is only 3 hours. A monoclonal capture antibody specific for Mouse IL-2 has been coated onto the wells of the 96-well plate provided. Samples, including a standard of known Mouse IL-2 content, controls, and unknowns, are pipetted into these wells followed by an addition of a biotinylated polyclonal detector antibody. During the first incubation, Mouse IL-2 antigen binds simultaneously to the immobilized capture antibody and to the solution phase biotinylated detector antibody on a second site. After washing, a streptavidin-horseradish peroxidase (HRP) is added. This binds to the biotinylated detection antibody to complete the four–member sandwich. After a second incubation and washing to remove all unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of Mouse IL-2 present in the original specimen and the optical density can be read on a standard microplate reader.

Ready-to-Use Mouse IL-2 ELISA Kit Contains These Reagents, Buffers, and Supplies:
• Mouse IL-2 Antibody Coated 96-Well Plate
• Mouse IL-2 Standard
• Standard Diluent Buffer
• Mouse IL-2 Detection Antibody
• Streptavidin-HRP
• HRP Diluent
• Wash Buffer Concentrate (25X)
• Stabilized Chromogen, TMB
• Stop Solution
• Plate Covers
• Detailed protocol with validation tests

Convenient Kit Format
The kit comes with a capture antibody precoated to the plate, eliminating an overnight coating process. The plates are machine coated to provide low variability well-to-well. Furthermore, the plate comes as removable 8-well strips allowing you to run as few samples as you want.

Each ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Wet ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Colorimetric
<8pg/mL
50μL
Interleukin 2
16183
RUO - research use only
Plasma, Serum, Supernatants
3 hrs