Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-2 Monkey ELISA Kit DFS Item


Promotes T-cell growth

Manufacturer: invitrogen™  LSKPC0022

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KPC0022

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
For Use With (Application) Colorimetric
Precision <2pg/mL
Sample Type Serum, Supernatants
Regulatory Status RUO - research use only
Quantity 192 assays
Test Time 4 hrs
Gene ID (Entrez) 3558
Sample Size 50μL
Target Interleukin 2

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.