Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-1B Mouse ELISA Kit DFS Item


Important in immune response

Manufacturer: invitrogen™  LSKMC0011

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KMC0011

  • / Each
 

Description & Specifications

Specifications

Precision <7pg/mL
Regulatory Status RUO - research use only
Gene ID (Entrez) 16176
Test Time 2.75 hrs
Sample Type Plasma, Serum, Supernatants
Includes Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers.
Quantity 96 assays
For Use With (Application) Colorimetric
Target Interleukin 1β
Sample Size 50μL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.