Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IL-15 Human ELISA Kit DFS Item


Quantifies natural and recombinant human IL-15 in serum, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKHC0151

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHC0151

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Sample Type Serum, Supernatants
Sensitivity <10pg/mL
Sample Size 50μL
Gene ID (Entrez) 3600
Quantity 96 assays
For Use With (Application) Colorimetric Detection
Accession Number NM_000585.4, NM_172174.2, NM_172175.2, NP_000576.1, NP_751915.1
For Use With (Equipment) Microplate Reader
Target interleukin 15
Marker Gene IL-15
Incubation Time 2 hr.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.