missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IL-15 Human ELISA Kit DFS Item

Quantifies natural and recombinant human IL-15 in serum, buffered solution, or cell culture medium

$526.00 - $980.00

Specifications

Gene ID (Entrez) 3600
Sample Size 50μL
Sample Type Serum, Supernatants
Sensitivity <10pg/mL
Target interleukin 15
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KHC0152 Invitrogen™
LSKHC0152
192 assays Each for $980.00
KHC0151 Invitrogen™
LSKHC0151
96 assays Each for $526.00
Specifications

Description

IL-15 is a cytokine.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

3600
Serum, Supernatants
interleukin 15
50μL
<10pg/mL