Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IL-15 Human ELISA Kit DFS Item


Quantifies natural and recombinant human IL-15 in serum, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKHC0152

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHC0152

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Microplate Reader
Sensitivity <10pg/mL
Gene ID (Entrez) 3600
Sample Type Serum, Supernatants
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Sample Size 50μL
Quantity 192 assays
For Use With (Application) Colorimetric Detection
Target interleukin 15
Accession Number NM_000585.4, NM_172174.2, NM_172175.2, NP_000576.1, NP_751915.1
Incubation Time 2 hr.
Marker Gene IL-15

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.