Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-13 Human ELISA Kit DFS Item


A cytokine produced primarily by Th2 cells

Manufacturer: invitrogen™  LSKHC0132

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHC0132

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Colorimetric
Precision <6pg/mL
Sample Type Plasma, Serum, Supernatants
Target Interleukin 13
Gene ID (Entrez) 3596
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Quantity 192 assays
Regulatory Status RUO - research use only
Sample Size 50μL
Test Time 4 hrs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.