Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-12 Mouse ELISA Kit DFS Item


A lymphokine

Manufacturer: invitrogen™  LSKMC0121C

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KMC0121C

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Sample Size 50μL
Quantity 480 assays
Target Interleukin 12a
Precision <2pg/mL
Sample Type Serum, Supernatants
Gene ID (Entrez) 16159
Test Time 3 hrs
For Use With (Application) Colorimetric
Regulatory Status RUO - research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.