Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-12 Human ELISA Kit DFS Item

Quantifies natural and recombinant human IL-12 p35/p40 heterodimer (or IL-12p70)

$558.60 - $980.70

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 3592
Precision <2pg/mL
Regulatory Status RUO - research use only
Sample Size 50μL
View More Specs
Includes:  Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

KHC0121

DFS Item
 
invitrogen™
LSKHC0121
96 assays Each for $558.60

KHC0122

DFS Item
 
invitrogen™
LSKHC0122
192 assays Each for $980.70
Description & Specifications

Specifications

For Use With (Application) Colorimetric
Gene ID (Entrez) 3592
Precision <2pg/mL
Regulatory Status RUO - research use only
Sample Size 50μL
Sample Type Plasma, Serum, Supernatants
Target Interleukin 12A
Test Time 3 hrs

Detects the free p40 subunit in serum, plasma, buffered solution, or cell culture medium

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Shipping Condition: Wet ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.