Please login to your online account to display your discounted pricing

Invitrogen™ Novex IL-10 Mouse ELISA Kit DFS Item


A lymphokine involved in immune response

Manufacturer: invitrogen™  LSKMC0101

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KMC0101

  • / Each
 

Description & Specifications

Specifications

Precision <13pg/mL
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
For Use With (Application) Colorimetric
Quantity 96 assays
Sample Size 100μL
Test Time 3 hrs
Sample Type Serum, Supernatants
Regulatory Status RUO - research use only
Target Interleukin 10
Gene ID (Entrez) 16153

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.