Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IFN-γ Monkey ELISA Kit DFS Item


Quantifies natural and recombinant monkey IFN γ in serum, plasma, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKPC4021

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KPC4021

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3458
Quantity 96 assays
Marker Gene IFN-γ
Incubation Time 4 hr.
Sample Size 50μL
Accession Number NM_000619.2, NP_000610.2
Sample Type Plasma, Serum, Supernatants
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers.
For Use With (Application) Colorimetric Detection
Sensitivity <4pg/mL
Target interferon, gamma
For Use With (Equipment) Microplate Reader

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.