missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IFN-γ Monkey ELISA Kit DFS Item

Quantifies natural and recombinant monkey IFN γ in serum, plasma, buffered solution, or cell culture medium

$526.00 - $980.70

Specifications

Gene ID (Entrez) 3458
Sample Size 50μL
Sample Type Plasma, Serum, Supernatants
Sensitivity <4pg/mL
Target interferon, gamma
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KPC4021 Invitrogen™
LSKPC4021
96 assays Each for $526.00
KPC4022 Invitrogen™
LSKPC4022
192 assays Each for $980.70
Specifications

Description

IFN γ is a lymphokine produced by activated T and NK cells.

Cell Analysis, Cell Signaling, Cell Viability, Proliferation & Function, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

3458
Plasma, Serum, Supernatants
interferon, gamma
50μL
<4pg/mL