Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IFN-γ Monkey ELISA Kit DFS Item


Quantifies natural and recombinant monkey IFN γ in serum, plasma, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKPC4022

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KPC4022

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers. Store Kit at 2-8°C.
Sample Type Plasma, Serum, Supernatants
For Use With (Equipment) Microplate Reader
Accession Number NM_000619.2, NP_000610.2
Sensitivity <4pg/mL
Sample Size 50μL
Target interferon, gamma
Quantity 192 assays
For Use With (Application) Colorimetric Detection
Incubation Time 4 hr.
Marker Gene IFN-γ
Gene ID (Entrez) 3458

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.