Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ IFN-γ Human ELISA Kit DFS Item


Each ready-to-use kit ELISA kit is validated for sensitivity, specificity, precision, and lot-to-lot consistency.

Manufacturer: invitrogen™  LSKHC4021C

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHC4021C

  • / Each
 

Description & Specifications

Specifications

Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Quantity 480 assays
Accession Number NM_000619.2, NP_000610.2
For Use With (Application) Colorimetric Detection
Gene ID (Entrez) 3458
Incubation Time 2.75 hr.
Sample Type Serum, Supernatants
For Use With (Equipment) Microplate Reader
Sensitivity <4pg/mL
Sample Size 50μL
Target interferon, gamma
Marker Gene IFN-γ

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.