Please login to your online account to display your discounted pricing
Invitrogen™ Novex™ iBlot™ Western Detection Stacks DFS Item
Uses the iBlot Dry Blotting System to speed up Western detection
Manufacturer: invitrogen™  IB701002
 View all options for this product
Western detection reagents
Catalog No. IB701002
Description & Specifications
Product Type iBlot Western Detection Stack
Dimensions (L x W) 8 x 8cm
Reduce time needed for Western detection from overnight to typically less than 30 minutes.
  • Number of stacks: 10
  • Available in two sizes: Regular (13.5 x 8cm); Mini (8 x 8cm)
Not a complete kit
Proteins, Expression, Isolation and Analysis, Western Blotting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.