missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ iBlot™ 2 Transfer Stacks, PVDF, mini DFS Item

Consumable stacks with integrated PVDF transfer membranes for dry blotting of proteins

Supplier:  Invitrogen™ IB24002

 View more versions of this product

Catalog No. IB24002



Description

Description

Each stack contains a copper coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins.

  • Increased shelf life
  • Reduced and compact packaging
  • Complete compatibility with iBlot 2 Gel Transfer Device

PVDF Membrane Features

  • 0.2μm pore size, low fluorescence
  • Higher binding capacity than nitrocellulose
  • Preactivated and ready for use without any pretreatment with alcohols
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic interactions
  • Protein-binding capacity: 240μ/cm2
Transfer Stack Sizes
  • The regular size (13 x 8.3cm) of this stack is used for all Novex midi gels (or other midi gel types), two Novex mini gels (or other mini gel types), and E-PAGE™ 48 or 96 gels
  • The mini size (8 x 8cm) is used for all Novex mini gels (or other mini gel types)

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

8 cm
Transfer Stack
Store at room temperature.
PVDF
10 blots
8 cm
0.2 μm
Mini, 8 cm x 8 cm
iBlot™
Less waste, sustainable packaging, sustainable disposal
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.