Invitrogen™ Novex™ iBlot™ 2 Nitrocellulose Transfer Stacks DFS Item
Used to transfer proteins with the iBlot 2 Gel Transfer Device
$208.00 - $254.00
Specifications
Material Nitrocellulose, 0.2μm pore size
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
IB23001 Invitrogen™
LSIB23001
Regular, 13.5 x 8cm Each for $254.00
IB23002 Invitrogen™
LSIB23002
Mini, 8 x 8cm Each for $208.00
Specifications
Description
The iBlot 2 Transfer Stacks are consumable stacks with integrated nitrocellulose transfer membranes for dry blotting of proteins. Each stack contains a copper coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins.
  • Increased shelf life
  • Reduced and compact packaging
  • Complete compatibility with iBlot 2 Gel Transfer Device
  • The nitrocellulose membrane (0.2μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer
  • Membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Protein binding capacity is 209μg/cm2
Transfer Stack Sizes
  • The regular size (13 x 8.3cm) of this stack is used for all Novex midi gels (or other midi gel types), two Novex mini gels (or other mini gel types), and E-PAGE™ 48 or 96 gels
  • The mini size (8 x 8cm) is used for all Novex mini gels (or other mini gel types)
Proteins, Expression, Isolation and Analysis, Western Blotting
Description & Specifications
Specifications
Nitrocellulose, 0.2μm pore size