Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ G-CSF Human ELISA Kit DFS Item


Quantifies natural and recombinant human Granulocyte Colony Stimulating Factor (G-CSF) in serum, plasma, buffered solution, or cell culture medium

Manufacturer: invitrogen™  LSKHC2032

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHC2032

  • / Each
 

Description & Specifications

Specifications

Sample Type Plasma, Serum, Supernatants
Incubation Time 3 hr.
Sensitivity <20pg/mL
Target Granulocyte colony stimulating factor 3
Quantity 192 assays
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Sample Size 50μL
For Use With (Equipment) Microplate Reader
For Use With (Application) Colorimetric Detection
Marker Gene G-CSF
Gene ID (Entrez) 1440

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.