missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ G-CSF Human ELISA Kit DFS Item

Quantifies natural and recombinant human Granulocyte Colony Stimulating Factor (G-CSF) in serum, plasma, buffered solution, or cell culture medium

$526.00 - $980.00

Specifications

Gene ID (Entrez) 1440
Sample Size 50μL
Sample Type Plasma, Serum, Supernatants
Sensitivity <20pg/mL
Target Granulocyte colony stimulating factor 3
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KHC2031 Invitrogen™
LSKHC2031
96 assays Each for $526.00
KHC2032 Invitrogen™
LSKHC2032
192 assays Each for $980.00
Specifications

Description

G-CSF supports growth of hematopoietic colonies.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

1440
Plasma, Serum, Supernatants
Granulocyte colony stimulating factor 3
50μL
<20pg/mL