Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Empty Gel Cassette Combs, Mini DFS Item

Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)

$81.50 - $84.50

Specifications

For Use With (Equipment) Mini-size Empty Gel Cassettes
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Product Type Thickness (Metric) Price Quantity    

NC3010

DFS Item
 
invitrogen™
LSNC3010
10-well Empty Gel Cassette Comb 1.0mm Each of 25 for $83.75

NC3001

DFS Item
 
invitrogen™
LSNC3001
1-well Empty Gel Cassette Comb 1.0mm Each for $82.75

NC3003

DFS Item
 
invitrogen™
LSNC3003
2-well Empty Gel Cassette Comb 1.0mm Each for $84.50

NC3015

DFS Item
 
invitrogen™
LSNC3015
15-well Empty Gel Cassette Comb 1.0mm Each for $81.50

NC3502

DFS Item
 
invitrogen™
LSNC3502
2D-well Empty Gel Cassette Comb 1.5mm Each for $81.90

NC3515

DFS Item
 
invitrogen™
LSNC3515
15-well Empty Gel Cassette Comb 1.5mm Each for $83.75

NC3002

DFS Item
 
life technologies
LSNC3002
2D-well Empty Gel Cassette Comb 1.0mm Each for $82.75
Description & Specifications

Specifications

For Use With (Equipment) Mini-size Empty Gel Cassettes

These combs are for use when pouring special gel types and are available in two thicknesses (1.0mm and 1.5mm) and seven well formats (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).

Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Shipping condition: Room Temperature