Please login to your online account to display your discounted pricing
Invitrogen™ Novex™ Empty Gel Cassette Combs, Mini DFS Item
Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)
Manufacturer: invitrogen™  LSNC3502
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC3502
  • / Each
Description & Specifications
Specifications
Product Type 2D-well Empty Gel Cassette Comb
For Use With (Equipment) Mini-size Empty Gel Cassettes
Thickness (Metric) 1.5mm