Invitrogen™ Novex™ Empty Gel Cassette Combs, Mini DFS Item
Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)
Manufacturer:  Invitrogen™ LSNC3010
 View more versions of this product
Catalog No. NC3010
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Mini-size Empty Gel Cassettes
10-well Empty Gel Cassette Comb
1.0mm