Please login to your online account to display your discounted pricing

Novex™ DYNAL™ Dynabeads™ M-280 Sheep Anti-Mouse IgG DFS Item


2.8μm superparamagnetic beads with affinity purified polyclonal sheep anti-mouse IgG covalently bound to bead surface

Manufacturer: life technologies  LS11201D

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11201D

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity All species, except mouse
Storage Requirements Storage: 2°C to 8°C.
Concentration 10mg/mL
Shelf Life 36 months
Dye Type Dynabeads™ M-280
Quantity 2mL
Applications Antibody Purification, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Immunoprecipitation, Organelle Isolation, Phage Display, Protein and Peptide Purification, Protein Complex Isolation and Analysis, Protein Complex Pull-Down, Protein Sample Preparation and Protein Purification, Protein-Protein Interactions, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research
Includes 2mL Dynabeads M-280 Sheep anti-Mouse IgG (10mg beads/mL)
Accessories

36 months