missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ DryEase™ Mini-Gel Drying System Replacement Parts DFS Item

For use with DryEase™ Mini-Gel Drying System

$63.25 - $418.00

Specifications

For Use With (Equipment) DryEase™ Mini-Gel Drying System
View More Specs

Products 4
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
NI2100 View Documents Invitrogen™
LSNI2100
Clamp Each for $63.25
 
NI2300 View Documents Invitrogen™
LSNI2300
Base Each for $125.00
 
NI2380 View Documents Invitrogen™
LSNI2380
Frame Each for $215.00
 
NI2387 View Documents Invitrogen™
LSNI2387
Drying System Each for $418.00
 
Description

Description

Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Specifications

DryEase™ Mini-Gel Drying System

For Research Use Only. Not for use in diagnostic procedures.