missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Caspase-8 Colorimetric Protease Assay Kit DFS Item

Designed to measure activity of Caspase-8

$340.00 - $904.00

Specifications

Gene ID (Entrez) 841
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Lysis buffer, substrate, reaction buffer, reducing agent, and complete protocol


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
KHZ0062 Invitrogen™
LSKHZ0062
pNA 200 tests Each for $904.00
KHZ0061 Invitrogen™
LSKHZ0061
pNA 25 tests Each for $340.00
Specifications

Description

The kit includes optimized buffers and substrate, IETD-AFC, composed of 7-amino-4-trifluoromethylcoumarin (AFC) and a synthetic tetrapeptide.

  • Includes 96-well plate, cuvettes

Apoptosis, Apoptosis Related Factors, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Dry ice

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

841