Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Caspase-8 Colorimetric Protease Assay Kit DFS Item


Designed to measure activity of Caspase-8

Manufacturer: invitrogen™  LSKHZ0062

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. KHZ0062

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 841
Quantity 200 tests
Conjugate pNA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.