Invitrogen™ Novex™ Bolt™ Mini Gel Tank Replacement Parts DFS Item
For the Bolt Mini Gel Tank
$33.73 - $174.30
Specifications
For Use With (Equipment) Bolt
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
B4478641 Invitrogen™
LSB4478641
Gel Runner Tank Each for $174.00
B4478592 Life Technologies
LSB4478592
Cassette Clamp, Right Each for $116.55
B4478591 Life Technologies
LSB4478591
Tank Lid Each for $174.30
B4478593 Life Technologies
LSB4478593
Cassette Clamp, Left Each for $148.05
B4478640 Life Technologies
LSB4478640
Tank Base Each for $33.73
Specifications
Description
The Bolt Tank Lid fits the top of the Gel Runner Tank. The Cassette Clamps (right and left) lock and seal the gel cassettes within the Gel Runner Tank, and the orientation is determined when facing the front of the gel tank. The Bolt Tank Base holds the Gel Runner Tank. The replacement Gel Runner Tank does not include tank lid, tank base, or cassette clamps.
The Bolt Mini Gel Tank is compatible with Bolt Bis-Tris Plus gels, and can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
Bolt Bis-Tris Plus Gels
  • Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
  • Similar to NuPAGE™ Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
  • Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial; also use Bolt gels to obtain optimal results for day-to-day protein separation needs
  • Bolt Bis-Tris Plus gels come in four gel types and multiple well formats
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Bolt