Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ 4X Bolt™ LDS Sample Buffer DFS Item


Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats

Manufacturer: invitrogen™  LSB0007

 View more versions of this product

lithium dodecyl sulfate (LDS) at pH 8.4.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. B0007

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 4˘ to 25°C.
Quantity 10mL
Concentration 4X
Formulation contains lithium dodecyl sulfate (LDS) at pH 8.4.

Novex™ Bolt™ buffers and sample reducing agents are optimized for use with Bolt Bis-Tris Plus gels, and are available in a variety of formats.

About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.

Bolt™ gel run times may vary depending upon the type of running buffer used.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.