MilliporeSigma™ Novagen™ ProteoExtract™ Glycopeptide Enrichment Kit
Designed to capture high-mannose, hybrid, and complex type glycosylated peptides from complex protein mixtures
Manufacturer:  MilliporeSigma™ 721033
Catalog No. 721033
Description
Description
  • Kit uses proprietary ZIC™ Glycocapture Resin
  • Non-specific and nonselective, without preference for particular glycan structures and without loss of distinct glycans
  • Directly compatible with downstream analysis by MALDI and ESI-MS
  • Provides sufficient reagents for 50 enrichment reactions
  • 10μL sample with total protein concentration 2 to 4mg/mL
Specifications
Specifications
1kit
Glycopeptide Enrichment Kit