EMD Millipore™ Novagen™ BLR(DE3) Competent Cells
For production of protein from target genes cloned in pET vectors by induction with IPTG
$114.00 - $228.77
Specifications
Type BLR(DE3) Competent Cells
Storage Requirements ≤ -70°C
View More Specs
Disclaimers:
For Research Use Only
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
690533 EMD Millipore™
690533
0.4mL Each for $114.00
690534 EMD Millipore™
690534
1mL Each for $228.77
Specifications
Description
  • recA- derivative of BL21
  • Improves plasmid monomer yields
  • Helps stabilize target plasmids containing repetitive sequences or whose products may cause loss of DE3 prophage (1, 2)
Description & Specifications
Specifications
BLR(DE3) Competent Cells
≤ -70°C