Please login to your online account to display your discounted pricing
EMD Millipore™ Novagen™ B834pLysS Competent Cells
For production of protein from target genes cloned in pET vectors by induction with IPTG
$144.00 - $241.00
Specifications
Type B834pLysS Competent Cells
Storage Requirements ≤ -70°C
View More Specs
 Disclaimers
For Research Use Only
Products
Catalog Number Mfr. No. Quantity Price Quantity    
690423
emd millipore™
690423
0.4mL Each for $144.00
690424
emd millipore™
690424
1mL Each for $241.00
Description & Specifications
Specifications
Type B834pLysS Competent Cells
Storage Requirements ≤ -70°C
  • B834 is parental strain for BL21 (1)
  • Protease-deficient hosts are methionine auxotrophs
  • Allows high specific-activity labeling of target proteins with methionine and selenomethionine for crystallography
  • pLysS strains express T7 lysozyme
  • Suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability