missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Notch2 Rat anti-Human, Mouse, PE, Clone: 16F11, eBioscience™

Rat Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen Notch2
Clone 16F11
Host Species Rat
Species Reactivity Human, Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010863 Life Technologies
LS12578680
25μg Each for $158.05
5010864 Life Technologies
LS12578682
100μg Each for $404.55
Specifications

Description

This 16F11 monoclonal antibody reacts with the extracellular domain of human and mouse Notch2, one of four members of the Notch family of receptors. Notch receptors are 300kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by γ-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription.

Notch2 is expressed on thymocytes (i.e., DN1 and DN2 subsets), activated peripheral T cells, and bone marrow. Moreover, expression of Notch2 has been reported to be altered in various cancers and lymphomas. In addition to its role in stem cell hematopoiesis, Notch2 plays a pivotal role in marginal zone B cell development. Notch2 also interacts with phosphorylated CREB1 to induce granzyme B transcription in cytotoxic T cells.
Description & Specifications

Specifications

Notch2
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
16F11
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer