missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Notch-1 Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Notch-1 Ab-1, Mouse Monoclonal Antibody.

$315.63 - $1,395.37

Specifications

Antigen Notch-1 Ab-1
Clone A6
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1339P Thermo Scientific™ Lab Vision™
MS1339P
200μg/mL 1mL Each for $1,319.96
MS1339P0 Thermo Scientific™ Lab Vision™
MS1339P0
200μg/mL 100μL Each for $315.63
MS1339P1 Thermo Scientific™ Lab Vision™
MS1339P1
200μg/mL 500μL Each for $791.31
MS1339P1ABX Thermo Scientific™ Lab Vision™
MS1339P1ABX
1mg/mL 100μL Each for $873.41
MS1339PABX Thermo Scientific™ Lab Vision™
MS1339PABX
1mg/mL 200μL Each for $1,395.37
Specifications

Description

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Notch-1 Ab-1, Mouse Monoclonal Antibody.

The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals, which include notch-1. notch-2, notch-3, and notch-4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins δ and Serrate have been identified as ligands of notch-1. Mature notch proteins are heterodimeric receptors derived from the cleavage of notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm). Alteration of notch-1 signaling or expression may contribute to tumorigenesis.

Host Species: Mouse

Clone: A6

Isotype: IgG2b

Species Reactivity: Human and Mouse. Others not known.

Epitope: Ligand binding region

Immunogen: Recombinant protein encoding the ligand binding region of human Notch-1

Molecular Weight: 290kDa

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Not verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Notch-1 Ab-1
Flow Cytometry
Unlabeled
Concentrated
RUO
Primary
A6
Monoclonal
Human,Mouse,Human,Mouse
Murine
Neuroscience
IgG2b