missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nortropine, TCI America™

$30.56 - $125.28

Chemical Identifiers

CAS 538-09-0
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00047140
InChI Key YYMCYJLIYNNOMK-MEKDEQNOSA-N
Synonym 8-Azabicyclo[3.2.1]octan-3-ol
PubChem CID 73480
IUPAC Name (1R,5S)-8-azabicyclo[3.2.1]octan-3-ol
SMILES C1CC2CC(CC1N2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Qty  
N084325G missing translation for 'viewDocuments' TCI America
N084325G
25g Each for $125.28
 
N08435G missing translation for 'viewDocuments' TCI America
N08435G
5g Each for $30.56
 
Specifications

Chemical Identifiers

538-09-0
127.187
YYMCYJLIYNNOMK-MEKDEQNOSA-N
73480
C1CC2CC(CC1N2)O
C7H13NO
MFCD00047140
8-Azabicyclo[3.2.1]octan-3-ol
(1R,5S)-8-azabicyclo[3.2.1]octan-3-ol

Specifications

Undesignated
538-09-0
MFCD00047140
YYMCYJLIYNNOMK-MEKDEQNOSA-N
(1R,5S)-8-azabicyclo[3.2.1]octan-3-ol
73480
Crystal-Powder at 20°C
Nortropine
25g
C7H13NO
8-Azabicyclo[3.2.1]octan-3-ol
C1CC2CC(CC1N2)O
127.187
127.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : RD0700000

TSCA : No