Honeywell™ North™ Water-Jel Emergency Burn Care
Honeywell North™ Water-Jel Emergency Burn Care products provide quick relief in an emergency. They are ideal for treating injuries to the inner arm, foot instep or other areas where a smaller dressing would be suited.
Manufacturer: Honeywell™ SC06643
Description
Ideal first aid treatment for burns, providing emergency care until victim receives medical attention. Biodegradable, bacteriostatic and water soluble, immediately cools skin, relieves pain, and protects burn against contamination. Easy to store, transport and use—no need for water source or special training.
Burn Dressings
- Sterile bacteriostatic dressings relieve pain of small burns without adhering to skin
- Individually wrapped in convenient foil packs
- Provides all the benefits of sterile burn dressings while safely allowing access to nasal passages, mouth, and eyes
- 12 × 16 in. (30 × 41cm) mask comes in individual pouch
- Water soluble gel cools for fast pain relief
- Comes in dispenser pack with 25 individually wrapped foil packets, each 0.13 oz. (3.5g)
Specifications
Ideal for the inner arm, foot instep or other areas where this size dressing would be suited, protects burn against contamination | |
3.8mL (0.13 oz.) | |
Burn Treatment |