North W-Series Safety Eyewear
Narrower across the front, shorter at the temple, and slimmer in the nose bridge
$83.82 - $86.77
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003 standard.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity  
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity  
17-377-429 North Safety Products
W102*
Brown Frosted Espresso Pack of 10 Each for $83.82
17-377-430 North Safety Products
W103*
Clear Frosted Silver Pack of 10 Each for $86.77
Specifications
Description
Smaller sizing, modern designs, and updated styles promote compliance.
  • Protective wraparound design with 8- and 9-base curve lenses for the most secure, protective fit
  • Smooth temples with optical curve for all-day wear
  • W100 Series features adjustable nosepiece for added comfort
  • Available with scratch-resistant hardcoat or Fog-Ban™ antifog coating
Compliance
Meets ANSI™ Z87.1-2003 standard.
Description & Specifications
Specifications
Meets ANSI Z87.1-2003 standard.