Honeywell™ North™ V-Maxx Goggles
Direct- and indirect-vented models with Fog-Ban™ antifog, antiscratch coating for durability
Manufacturer:  Honeywell™ 11250800
 View more versions of this product
Catalog No. 19-065-516
Description
Description
  • Comfortable protection against impact, dust, mist and chemical splash
  • Wraparound design provides maximum coverage and 180° field of vision
  • Soft rubber construction ensures secure, comfortable fit
  • Large enough to be worn over prescription glasses
  • Red headband for impact-resistant vented models; black headband for chemical-splash indirect vented models
Red headband for impact-resistant vented models; black headband for chemical-splash indirect vented models
Order Info
Peel-off goggle covers are available; contact your Fisher Scientific Customer Service Representative.
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
ANSI Z87.1-2010, CSA Z94.3
Red
Elastic
Clear
Polycarbonate
Safety
Direct venting, closed
Fog-Ban Anti-fog
Anti-fog
Impact Resistant Goggles