Honeywell™ North Trawler King™ PVC Gloves
Chemically resistant to oils, greases, caustics, acids and alcohols
Manufacturer:  Honeywell™ 850FWG10XL
 View more versions of this product
Catalog No. 19-150-1104
Description
Description
  • Textured finish on palm and fingers
  • Multi-dipped construction
  • Pinked cuff
  • Interlock knit liner
  • Supported
  • Color: Green
Specifications
Specifications
PVC Nitrile blend
Gauntlet
construction, handling Oils and Greases, mining, refining, Petrochemical and fishing
PVC (Polyvinyl Chloride), Nitrile
ANSI/ISEA 105, ASTM D 3389-05, ANSI/ISEA 105, ASTM F 1790-97, EN 374, EN 388, EN 420, OSHA 29 CFR 1915.157
14 in.
Rough
Hand-specific
Jersey
X-Large
Green
35.5cm
Documents