Please login to your online account to display your discounted pricing
Sperian™ Survivair™ Opti-Fit™ APR Respirator
Compatible with S-Series cartridges and filters
$142.08 - $146.59
Specifications
Cartridge Type S Series and T Series Cartridge
Product Type Air Purifying Respirator
For Use With (Application) Recommended industries/use : agriculture, chemical, construction, fire protection, food services, forestry, government, law enforcement, manufacturing, medical, military, mining, municipal services, nuclear, oil and gas, pharmaceutical, steel and metals,
Material (Lens) Polycarbonate
Suspension Type 5 point head strap
View More Specs
Products
Catalog Number Mfr. No. Connection Type For Use With (Equipment) Material Size Price Quantity    
19-065-188A
north safety products
752000
Threaded Survivair™ s series cartridge Silicone Small Each for $142.08
19-065-188C
north safety products
772000
Threaded Survivair™ s series cartridge Silicone Large Each for $146.59
Description & Specifications
Specifications
Cartridge Type S Series and T Series Cartridge
Product Type Air Purifying Respirator
For Use With (Application) Recommended industries/use : agriculture, chemical, construction, fire protection, food services, forestry, government, law enforcement, manufacturing, medical, military, mining, municipal services, nuclear, oil and gas, pharmaceutical, steel and metals,
Material (Lens) Polycarbonate
Suspension Type 5 point head strap
Full-face APR respirator provides optimum fit, comfort and superior optics
  • Head harness: 5-point strap
  • Wide viewing area offers distortion free visibility and optical clarity
  • Silicone skirt provides exceptional durability and comfort
  • All straps are attached to rim of facepiece, rather than the silicone skirt, to reduce pressure points and distortion of the sealing surface when tightened
  • Nose cup prevents fogging with optional kits for customized sizing (comes standard with medium nose cup); optional clear nose cup kits available
  • Replaceable, high-impact resistant polycarbonate lens treated with anti-scratch coating
  • Molded in speaking diaphragm provides improved audio transmission for communication
  • Available in three sizes for wide range of fit
  • Hazards: Airborne Particulates, Biohazard, Chemical, Contamination, Extreme Environment, Gas, Vapors, Smoke
Applications:
Agriculture, Chemical, Construction, Fire Protection, Food Services, Forestry, Government, Law Enforcement, Manufacturing, Medical, Military, Mining, Municipal Services, Nuclear, Oil and Gas, Pharmaceutical, Steel and Metals, Transportation, Utilities, Welding.
NIOSH 42 CFR 84.
Free of defects in materials and workmanship for five years (for the skirt) and one year (for all other components) from date of purchase.