Honeywell™ North™ Sparkguard Headgear
Basic, simple-to-use headgear for all applications. A protected mounting system helps eliminate fluids from entering the backside of the faceshield, while an unprotected mounting system offers quick visor replacement. Available with the SmoothLok ratchet and Protected Fit systems.
Manufacturer:  Honeywell™ KHG5001
Catalog No. 29-635-2497
Description
Description
Patented non-slip ratchet system is easy to use and offers precise size adjustments. The SmoothLok ratchet is available on the KHG Series headgear for quick, easy and perfect fit. The 5000 Series comes with Protected Fit holding system for faceshield to help reinforce splash protection to inside of the faceshield.
  • Accepts a wide range of faceshield styles
  • Large sparkguard provides excellent protection
  • Cross strap adjusts to allow an exceptional fit
  • Full floating mount sparkguard provides comfortable wear even with heavy faceshield and continuous use
  • Meets ANSI Z87.1-2010
Specifications
Specifications
Black
SmoothLok ratchet adjustment
Headgear
Faceshield Visor
Plastic
Large