Honeywell™ North™ Qualitative Fit-Test Kits
Qualitative fit-testing kits offer a comprehensive, portable package for fit testing employees in compliance with OSHA regulations
Manufacturer:  Honeywell™ 770040
Catalog No. 17-622-19
Description
Description
Bitrex™ fit-testing kit is ideal for testing particulate-removing respirators, including filtering facepieces.
Irritant Smoke Fit-Testing Kit includes:
  • Training video
  • Six fit-test smoke tubes, bulb, and breaker
  • Instructions and wall chart
  • 20 refresher wipe pads to clean perspiration and body oils from facepiece between scheduled cleanings; certificates; wallet versions of certificates and qualitative fit-test reports
Bitrex Fit-Testing Kit includes:
  • Bitrex solution
  • Instructions and wall chart
  • 20 refresher wipe pads to clean perspiration and body oils from facepiece between scheduled cleanings; certificates; wallet versions of certificates and qualitative fit-test reports
Order Info
All components come in sturdy, plastic carrying case measuring 9.5L x 13.5W x 6 in.H (24.1 x 34.3 x 15.2cm).
Compliance
Accepted by OSHA for fit testing under 29CFR1910.34, Appendix A.
Specifications
Specifications
OSHA
Box of 6 fit test irritant smoke tubes, smoke tube bulb, smoke tube breaker, respirator refresher wipe pads (20), training DVD, respirator fitting guide wall chart, certificates of training and fit testing (20), wallet versions of training and fit testing certificates (20), requests for fit testing (4), requests for respirator use training (4)
1 Ea.
For quantitative and qualitative fit testing
Irritant Smoke Fit Testing Kit